missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperidine, ≥99.5%, Honeywell™
£89.97
Chemical Identifiers
CAS | 110-89-4 |
---|---|
Molecular Formula | C5H11N |
Molecular Weight (g/mol) | 85.15 |
MDL Number | MFCD00005979 |
InChI Key | NQRYJNQNLNOLGT-UHFFFAOYSA-N |
Synonym | hexahydropyridine, azacyclohexane, cyclopentimine, cypentil, hexazane, pentamethyleneimine, piperidin, pentamethylenimine, perhydropyridine, pentamethyleneamine |
PubChem CID | 8082 |
ChEBI | CHEBI:18049 |
IUPAC Name | piperidine |
SMILES | C1CCNCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15675860
|
Honeywell Burdick & Jackson
571261-200ML |
200 mL | N/A | ||||||
Description
Chemical Identifiers
110-89-4 | |
85.15 | |
NQRYJNQNLNOLGT-UHFFFAOYSA-N | |
8082 | |
piperidine |
C5H11N | |
MFCD00005979 | |
hexahydropyridine, azacyclohexane, cyclopentimine, cypentil, hexazane, pentamethyleneimine, piperidin, pentamethylenimine, perhydropyridine, pentamethyleneamine | |
CHEBI:18049 | |
C1CCNCC1 |
Specifications
110-89-4 | |
16°C (60.8°F) | |
MFCD00005979 | |
102438 | |
NQRYJNQNLNOLGT-UHFFFAOYSA-N | |
piperidine | |
8082 | |
85.15g/mol | |
Piperidine |
0.862g/mL at 20°C | |
C5H11N | |
UN2401 | |
hexahydropyridine, azacyclohexane, cyclopentimine, cypentil, hexazane, pentamethyleneimine, piperidin, pentamethylenimine, perhydropyridine, pentamethyleneamine | |
C1CCNCC1 | |
85.15 | |
CHEBI:18049 | |
≥99.5% |
Safety and Handling
P210-P280-P304 + P340 + P310-P305 + P351 + P338-P370 + P378-P403 + P235
missing translation for 'dotInformation' : 3(8)/PGI
missing translation for 'rtecsNumber' : 203-813-0
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title