missing translation for 'onlineSavingsMsg'
Learn More

Piperidine, ≥99.5%, Honeywell™

Product Code. 15675860
Click to view available options
Quantity:
200 mL
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Chemical Identifiers

CAS 110-89-4
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD00005979
InChI Key NQRYJNQNLNOLGT-UHFFFAOYSA-N
Synonym hexahydropyridine, azacyclohexane, cyclopentimine, cypentil, hexazane, pentamethyleneimine, piperidin, pentamethylenimine, perhydropyridine, pentamethyleneamine
PubChem CID 8082
ChEBI CHEBI:18049
IUPAC Name piperidine
SMILES C1CCNCC1

Specifications

Density 0.862g/mL at 20°C
Flash Point 16°C (60.8°F)
Quantity 200 mL
UN Number UN2401
Beilstein 102438
Formula Weight 85.15g/mol
Percent Purity ≥99.5%
Chemical Name or Material Piperidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.