missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric acid, ACS Reagent, ≥85 wt. % in H2O, Honeywell Fluka™
ACS Reagent,≥85 wt. % in H2O
£28.53 - £145.72
Chemical Identifiers
CAS | 7664-38-2, 7732-18-5 |
---|---|
Molecular Formula | H3O4P |
Molecular Weight (g/mol) | 97.994 |
MDL Number | MFCD00011340 |
InChI Key | NBIIXXVUZAFLBC-UHFFFAOYSA-N |
Synonym | orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico |
PubChem CID | 1004 |
ChEBI | CHEBI:26078 |
IUPAC Name | phosphoric acid |
SMILES | OP(=O)(O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15655530
|
Honeywell-Fluka
438081-500ML |
500 mL | Poly Bottle | N/A | |||||
15635530
|
Honeywell-Fluka
438081-2.5L |
2.5 L | Poly Bottle | N/A | |||||
15665530
|
Honeywell-Fluka
438081-6X500ML |
6 x 500 mL | Poly Bottle | N/A | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7664-38-2, 7732-18-5 | |
97.994 | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
1004 | |
phosphoric acid |
H3O4P | |
MFCD00011340 | |
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
CHEBI:26078 | |
OP(=O)(O)O |
Specifications
7664-38-2, 7732-18-5 | |
158°C | |
<90.0000%, <20.0000% | |
H3O4P | |
MFCD00011340 | |
1921286 | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
phosphoric acid | |
1004 | |
98 | |
ACS Reagent |
∼40°C (literature) | |
≥70.0000%, ≥10.0000% | |
1.685 g/mL at 25 °C(lit.) | |
H3PO4 | |
UN1805 | |
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
OP(=O)(O)O | |
97.994 | |
CHEBI:26078 | |
>85% | |
Phosphoric acid, ACS Reagent, ≥85 wt.% in H2O |
Safety and Handling
missing translation for 'shelfLife' : 900 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Phosphoric acid solution
missing translation for 'einecsNumber' : 231-633-2
Recommended Storage : Room Temp
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Phosphoric acid
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Orthophosphoric acid
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title