missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric acid, ACS Reagent, ≥85 wt. % in H2O, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
2.5 L
6 x 500 mL
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Melting Point | ∼40°C (literature) |
Boiling Point | 158°C |
CAS Min % | ≥70.0000%, ≥10.0000% |
CAS Max % | <90.0000%, <20.0000% |
Density | 1.685 g/mL at 25 °C(lit.) |
Packaging | Poly Bottle |
Quantity | 2.5 L |
Linear Formula | H3PO4 |
UN Number | UN1805 |
Beilstein | 1921286 |
Show More |
Safety and Handling
Product Identifier
- Phosphoric acid
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Orthophosphoric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction