missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol Red sodium salt, 0.02% w/v aq. soln., Thermo Scientific Chemicals
POA - £45.83
Chemical Identifiers
CAS | 34487-61-1 |
---|---|
Molecular Formula | C19H13NaO5S |
Molecular Weight (g/mol) | 376.358 |
MDL Number | MFCD00066901 |
InChI Key | HKHYOKBQJILTEI-UHFFFAOYSA-M |
PubChem CID | 23686673 |
IUPAC Name | sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate |
SMILES | C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11308977
|
Thermo Scientific Alfa Aesar
038702.AE |
100 mL |
£22.61
100mL |
In Stock
Log in to see stock. |
|||||
11410161
|
Thermo Scientific Alfa Aesar
038702.AP |
500 mL |
£45.83
500mL |
In Stock
Log in to see stock. |
|||||
11318977
|
Thermo Scientific Alfa Aesar
038702.M0 |
6 x 100 mL | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
34487-61-1 | |
376.358 | |
HKHYOKBQJILTEI-UHFFFAOYSA-M | |
sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate |
C19H13NaO5S | |
MFCD00066901 | |
23686673 | |
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+] |
Specifications
34487-61-1 | |
C19H13NaO5S | |
HKHYOKBQJILTEI-UHFFFAOYSA-M | |
sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate | |
23686673 | |
0.972 g/mL | |
Phenol Red sodium salt |
0.02% w/v aq. soln. | |
MFCD00066901 | |
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+] | |
376.358 | |
376.37 | |
Liquid |
Safety and Handling
missing translation for 'einecsNumber' : 252-057-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only