missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red sodium salt, 0.02% w/v aq. soln., Thermo Scientific Chemicals

Product Code. 11318977
Click to view available options
Quantity:
100 mL
500 mL
6 x 100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34487-61-1
Molecular Formula C19H13NaO5S
Molecular Weight (g/mol) 376.358
MDL Number MFCD00066901
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
PubChem CID 23686673
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]

Specifications

Concentration 0.02% w/v aq. soln.
Formula Weight 376.37
Density 0.972 g/mL
Quantity 6 x 100 mL
Physical Form Liquid
Chemical Name or Material Phenol Red sodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'