Learn More
Thermo Scientific Chemicals Penicillin V potassium salt, 94+%, Thermo Scientific Chemicals
£31.86 - £84.40
Chemical Identifiers
CAS | 132-98-9 |
---|---|
Molecular Formula | C16H17KN2O5S |
Molecular Weight (g/mol) | 388.48 |
MDL Number | MFCD00051771 |
InChI Key | HCTVWSOKIJULET-LQDWTQKMSA-M |
PubChem CID | 131673927 |
SMILES | [K+].CC1(C)S[C@@H]2[C@H](NC(=O)COC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15354998
|
Thermo Scientific Chemicals
455230050 |
5 g |
£31.86
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15364998
|
Thermo Scientific Chemicals
455230250 |
25 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
132-98-9 | |
388.48 | |
HCTVWSOKIJULET-LQDWTQKMSA-M | |
[K+].CC1(C)S[C@@H]2[C@H](NC(=O)COC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O |
C16H17KN2O5S | |
MFCD00051771 | |
131673927 |
Specifications
Penicillin V Potassium Salt | |
197.0°C to 202.0°C | |
+220° to +235° (20°C, 589nm) (c=1, H2O) | |
MFCD00051771 | |
HCTVWSOKIJULET-LQDWTQKMSA-M | |
(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenoxyacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;potassium | |
131673927 | |
≥98.5% (HPLC) |
132-98-9 | |
Authentic | |
C16H17KN2O5S | |
Solubility in water: soluble. | |
[K+].CC1(C)S[C@@H]2[C@H](NC(=O)COC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O | |
388.48 | |
388.48 | |
Penicillin V potassium salt |
Safety and Handling
GHS H Statement Harmful if swallowed. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement Wear eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Danger
- Penicillin V potassium salt
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
RUO – Research Use Only