missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Penicillin V potassium salt, 94+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Type | Penicillin V Potassium Salt |
Melting Point | 197.0°C to 202.0°C |
Infrared Spectrum | Authentic |
Specific Rotation | +220° to +235° (20°C, 589nm) (c=1, H2O) |
Quantity | 25 g |
Solubility Information | Solubility in water: soluble. |
IUPAC Name | potassium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenoxyacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate |
Formula Weight | 388.48 |
Percent Purity | ≥98.5% (HPLC) |
Chemical Name or Material | Penicillin V potassium salt |
Safety and Handling
Product Identifier
- Penicillin V potassium salt
- Danger
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'