missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vici Valco Instruments™ PEEK Tubing
Color-coded PEEK tubing strong enough for continuous use at HPLC pressure without swelling or bursting. Vici Valco Instruments™ PEEK Tubing is not affected by halide salts, high-strength buffers, or other aggressive mobile phases that corrode stainless steel.
£110.24 - £111.30
Specifications
Type | Tubing |
---|---|
For Use With (Application) | Chromatography |
Diameter (English) Outer | 1/16 in. |
Material | PEEK |
Max. Operating Temperature (Metric) | 100°C |
Product Code | Brand | Color | Diameter (Metric) Inner | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Diameter (Metric) Inner | Price | Quantity & Availability | ||||
13173563
|
Vici Valco Instruments™
724JRT5999M3 |
Red | 0.13 mm |
£111.30
3m |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
13113573
|
Vici Valco Instruments™
724JRT6003M3 |
Green | 0.75 mm |
£110.24
3m |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
Description
- Color-coded based on size (interior diameter) and length
- Strong enough to withstand continuous use at HPLC pressure without swelling or bursting
- Not affected by halide salts, high strength buffers, or other aggressive mobile phases that corrode stainless steel
- Polymer surface will not leach metal ions into the eluent
- Polymer surface will not extract metal-sensitive components from the sample
- Note: dichloromethane, THF, and DMSO may cause swelling in PEEK, and concentrated nitric and sulphuric acid will attack PEEK.
Specifications
Tubing | |
1/16 in. | |
100°C |
Chromatography | |
PEEK |