Learn More
Vici Valco Instruments™ PEEK Tubing
Color-coded PEEK tubing strong enough for continuous use at HPLC pressure without swelling or bursting. Vici Valco Instruments™ PEEK Tubing is not affected by halide salts, high-strength buffers, or other aggressive mobile phases that corrode stainless steel.
Brand: Vici Valco Instruments™ 724JRT5999M3
Code : 16
Additional Details : Weight : 0.01080kg
Description
- Color-coded based on size (interior diameter) and length
- Strong enough to withstand continuous use at HPLC pressure without swelling or bursting
- Not affected by halide salts, high strength buffers, or other aggressive mobile phases that corrode stainless steel
- Polymer surface will not leach metal ions into the eluent
- Polymer surface will not extract metal-sensitive components from the sample
- Note: dichloromethane, THF, and DMSO may cause swelling in PEEK, and concentrated nitric and sulphuric acid will attack PEEK.
Specifications
Tubing | |
Chromatography | |
0.13 mm | |
PEEK |
Red | |
1/16 in. | |
3 m | |
100°C |