missing translation for 'onlineSavingsMsg'
Learn More

Oxalyl bromide, 98%, Thermo Scientific Chemicals

£115.40 - £1191.80

Chemical Identifiers

CAS 15219-34-8
Molecular Formula C2Br2O2
Molecular Weight (g/mol) 215.83
MDL Number MFCD00000113
InChI Key JAZLVNXWYDFQFE-UHFFFAOYSA-N
Synonym oxalyl bromide, ethanedioyl dibromide, ethanedioyl bromide, oxalic dibromide, brcocobr, oxalyl bromide solution, ethanedioyl dibromide solution, oxalyl bromide solution, 2.0 m in methylene chloride
PubChem CID 84841
IUPAC Name oxalyl dibromide
SMILES BrC(=O)C(Br)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10112291
View Documents
Thermo Scientific Acros
150890250
25 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10662762
View Documents
Thermo Scientific Acros
150891000
100 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10155773
View Documents
Thermo Scientific Acros
150895000
500 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
Specifications

Chemical Identifiers

15219-34-8
215.83
JAZLVNXWYDFQFE-UHFFFAOYSA-N
84841
BrC(=O)C(Br)=O
C2Br2O2
MFCD00000113
oxalyl bromide, ethanedioyl dibromide, ethanedioyl bromide, oxalic dibromide, brcocobr, oxalyl bromide solution, ethanedioyl dibromide solution, oxalyl bromide solution, 2.0 m in methylene chloride
oxalyl dibromide

Specifications

15219-34-8
2.3000g/mL
Authentic
C2Br2O2
1.5205 to 1.5225
01,767; 01,1137
oxalyl bromide, ethanedioyl dibromide, ethanedioyl bromide, oxalic dibromide, brcocobr, oxalyl bromide solution, ethanedioyl dibromide solution, oxalyl bromide solution, 2.0 m in methylene chloride
JAZLVNXWYDFQFE-UHFFFAOYSA-N
oxalyl dibromide
84841
98%
Oxalyl bromide
-19.0°C
102.0°C to 103.0°C (720.0 mmHg)
97.5% min. (Argentometry)
BrCOCOBr
MFCD00000113
2.3
Solubility in water: reacts with water.
BrC(=O)C(Br)=O
215.83
215.83
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 239-271-7

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.