missing translation for 'onlineSavingsMsg'
Learn More

Oxalyl bromide, 98%, Thermo Scientific Chemicals

Product Code. 10155773
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15219-34-8
Molecular Formula C2Br2O2
Molecular Weight (g/mol) 215.83
MDL Number MFCD00000113
InChI Key JAZLVNXWYDFQFE-UHFFFAOYSA-N
Synonym oxalyl bromide, ethanedioyl dibromide, ethanedioyl bromide, oxalic dibromide, brcocobr, oxalyl bromide solution, ethanedioyl dibromide solution, oxalyl bromide solution, 2.0 m in methylene chloride
PubChem CID 84841
IUPAC Name oxalyl dibromide
SMILES BrC(=O)C(Br)=O

Specifications

Melting Point -19.0°C
Density 2.3000g/mL
Boiling Point 102.0°C to 103.0°C (720.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula BrCOCOBr
Refractive Index 1.5205 to 1.5225
Quantity 500 g
Fieser 01,767; 01,1137
Specific Gravity 2.3
Solubility Information Solubility in water: reacts with water.
Formula Weight 215.83
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Oxalyl bromide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.