Learn More
Thermo Scientific™ Orion™ Cyanide Electrodes
Measure cyanide ions in aqueous solutions quickly, simply, accurately and economically with these cyanide electrodes.
Brand: Thermo Scientific™ 9406SC
Code : 59
Additional Details : Weight : 0.50000kg
Description
Measure cyanide ions in aqueous solutions quickly, simply and economically with Thermo Scientific™ Orion™ Cyanide Electrodes. No separate reference electrode is needed with this electrode, making it convenient to use with small sample volumes.
No separate reference electrode is needed with this electrode, making it convenient to use with small sample volumes.- Calibration Standards Catalog number: Consult instruction manual
- Required ISA Catalog number: 951011
Specifications
Screw Cap | |
Free-flowing liquid junction assures stable, drift-free potentials | |
Combination Ion Selective Electrode |
With screw cap connector | |
ISE Meters |