missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NUAK1 (ARK5), His Tag Recombinant Protein
Human NUAK1 (ARK5), His Tag Recombinant Protein
£437.00 - £6900.00
Specifications
Accession Number | NP_055655 |
---|---|
Gene Alias | NUAK1 (ARK5) |
KinaseFamily | CAMKL Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15438613
|
Invitrogen™
PR7569A |
100μg |
£1588.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10536573
|
Invitrogen™
PV4283 |
1mg |
£6900.00
1mg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11573617
|
Invitrogen™
PV4127 |
10 μg |
£437.00
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The overexpression of the NUAK1 (ARK5) kinase has been associated with tumor invasion and metastasis (colorectal neoplasm).
Specifications
NP_055655 | |
CAMKL Kinase Family | |
Recombinant | |
Baculovirus |
NUAK1 (ARK5) | |
Human | |
His Tag |