missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NUAK1 (ARK5), His Tag Recombinant Protein
Human NUAK1 (ARK5), His Tag Recombinant Protein
Brand: Invitrogen™ PV4283
Code : NEW
Additional Details : Weight : 0.00900kg
Description
The overexpression of the NUAK1 (ARK5) kinase has been associated with tumor invasion and metastasis (colorectal neoplasm).
Specifications
NP_055655 | |
0mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% TritonX-100, 2mM DTT, 50% Glycerol | |
1mg | |
RUO - research use only | |
NUAK1 (ARK5) | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
78.3kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CAMKL Kinase Family | |
Recombinant | |
Baculovirus |