missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Miscellaneous Microtome Section Waste Tray with Arm Rest, For HM 310, HM 315, HM325, and HM 335

Support and expand the functionality of Epredia™ 200 and 300 series of microtomes with Epredia™ Miscellaneous Microtome Accessories.

£236.00 - £1140.00

Specifications

For Use With (Equipment) Epredia 200/300 Series Microtomes
View More Specs

Products 2
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
12063089
View Documents
Epredia™
640280
Foot Pedal This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10871330
View Documents
Epredia™
760170
Magnifier This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Interlock plug replaces foot pedal connector
  • Storage unit can be removed and used separately; tray for specimens can be cooled by changeable inserts
  • Large-field magnifier attaches to facilitate knife alignment and approach for sectioning; magnification 2.5X
Specifications

Specifications

Epredia 200/300 Series Microtomes
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.