Learn More
Epredia™ Miscellaneous Microtome Section Waste Tray with Arm Rest, For HM 310, HM 315, HM325, and HM 335
Support and expand the functionality of Epredia™ 200 and 300 series of microtomes with Epredia™ Miscellaneous Microtome Accessories.
Brand: Epredia™ 760170
This item is not returnable.
View return policy
Description
- Interlock plug replaces foot pedal connector
- Storage unit can be removed and used separately; tray for specimens can be cooled by changeable inserts
- Large-field magnifier attaches to facilitate knife alignment and approach for sectioning; magnification 2.5X
Specifications
Large-field Magnifier | |
Magnifier | |
Epredia 200/300 Series Microtomes |