Learn More
Thermo Scientific Chemicals Methyl Red sodium salt
£23.52 - £109.55
Chemical Identifiers
CAS | 845-10-3 |
---|---|
Molecular Formula | C15H14N3NaO2 |
Molecular Weight (g/mol) | 291.286 |
MDL Number | MFCD00002426 |
InChI Key | GNTPCYMJCJNRQB-UHFFFAOYSA-M |
Synonym | C.I. 13020; Methyl Red, water soluble |
PubChem CID | 4465632 |
IUPAC Name | sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate |
SMILES | CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11438047
|
Thermo Scientific Chemicals
A17455.14 |
25 g |
£23.52
25g |
In Stock
Log in to see stock. |
|||||
11448047
|
Thermo Scientific Chemicals
A17455.22 |
100 g |
£35.48
100g |
In Stock
Log in to see stock. |
|||||
11458047
|
Thermo Scientific Chemicals
A17455.36 |
500 g |
£109.55
500g |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
Description
Methyl red is an indicator dye that turns red in acidic solutions. It is a pH indicator, turning red under pH 4.4, yellow over pH 6.2, and orange in between those values. It is used in microbiology in the Voges-Proskauer test to identify bacteria that produce stable acids through mixed acid fermentation of glucose. Methyl-red positive bacteria include Proteus vulgaris and Escherichia coli. Methyl-red negative bacteria include Enterobacter aerogenes and Serratia marcescens.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl red is an indicator dye that turns red in acidic solutions. It is a pH indicator, turning red under pH 4.4, yellow over pH 6.2, and orange in between those values. It is used in microbiology in the Voges-Proskauer test to identify bacteria that produce stable acids through mixed acid fermentation of glucose. Methyl-red positive bacteria include Proteus vulgaris and Escherichia coli. Methyl-red negative bacteria include Enterobacter aerogenes and Serratia marcescens.
Solubility
Soluble in water (800 mg/ml at 25°C), and slightly soluble in ethanol at (25°C).
Notes
Store at room temperature. Reactive with oxidizing agents.
Chemical Identifiers
845-10-3 | |
291.286 | |
GNTPCYMJCJNRQB-UHFFFAOYSA-M | |
4465632 | |
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+] |
C15H14N3NaO2 | |
MFCD00002426 | |
C.I. 13020; Methyl Red, water soluble | |
sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate |
Specifications
845-10-3 | |
MFCD00002426 | |
C.I. 13020; Methyl Red, water soluble | |
GNTPCYMJCJNRQB-UHFFFAOYSA-M | |
sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate | |
4465632 | |
Methyl Red sodium salt |
C15H14N3NaO2 | |
4065488 | |
Soluble in water (800mg/ml at 25°C),and slightly soluble in ethanol at (25°C). | |
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+] | |
291.286 | |
291.29 |
Safety and Handling
missing translation for 'einecsNumber' : 212-682-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only