missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Red sodium salt

Product Code. 11448047
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Methyl red is an indicator dye that turns red in acidic solutions. It is a pH indicator, turning red under pH 4.4, yellow over pH 6.2, and orange in between those values. It is used in microbiology in the Voges-Proskauer test to identify bacteria that produce stable acids through mixed acid fermentation of glucose. Methyl-red positive bacteria include Proteus vulgaris and Escherichia coli. Methyl-red negative bacteria include Enterobacter aerogenes and Serratia marcescens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl red is an indicator dye that turns red in acidic solutions. It is a pH indicator, turning red under pH 4.4, yellow over pH 6.2, and orange in between those values. It is used in microbiology in the Voges-Proskauer test to identify bacteria that produce stable acids through mixed acid fermentation of glucose. Methyl-red positive bacteria include Proteus vulgaris and Escherichia coli. Methyl-red negative bacteria include Enterobacter aerogenes and Serratia marcescens.

Solubility
Soluble in water (800 mg/ml at 25°C), and slightly soluble in ethanol at (25°C).

Notes
Store at room temperature. Reactive with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 845-10-3
Molecular Formula C15H14N3NaO2
Molecular Weight (g/mol) 291.286
MDL Number MFCD00002426
InChI Key GNTPCYMJCJNRQB-UHFFFAOYSA-M
Synonym C.I. 13020; Methyl Red, water soluble
PubChem CID 4465632
IUPAC Name sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]

Specifications

Beilstein 4065488
Solubility Information Soluble in water (800mg/ml at 25°C),and slightly soluble in ethanol at (25°C).
Formula Weight 291.29
Quantity 100 g
Chemical Name or Material Methyl Red sodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'