Learn More
Emma Safety Footwear Martino (Frontier 162) Safety Shoes
The executive safety shoe is made with the same care and  attention to detail associated with  high end luxury shoes. Brazilian 2 tone full grain leather combined with nubuck adds to the stylish look.
Brand:  Emma Safety Footwear MM16209041
Additional Details : Weight : 1.96900kg
 View more versions of this product
Product Code. 15928271
  • £237.44 / 1 pair
Estimated Shipment: 17-06-2024
to see stock.
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Thermoplastic Polyurethane (TPU) Outsole
  • Full grain leather construction
  • ESD certified
  • Chrome-free lining
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011, EN 12568:2010
Brown
Hydro-Tec™ Sanitized Silver & Wellness leather
41
Laces
Martino (Frontier 162)
Leather
Steel