missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilson™ MACROMAN™ Wall Mounts
These wall mounts provide safe and stable storage for your MACROMAN™ pipette controllers.
£56.30 - £84.35
Specifications
Color | Black |
---|---|
Type | Wall Mount |
For Use With (Application) | Storage |
For Use With (Equipment) | MACROMAN™ Pipette Controller |
Description
- Keeps pipette controllers away from busy benches and desktops
- Available in magnetic and screw varieties
- Accessory for MACROMAN pipette controllers
- Supplied individually
Specifications
Black | |
Wall Mount | |
Storage | |
MACROMAN™ Pipette Controller |