Learn More
Gilson™ MACROMAN™ Wall Mounts
These wall mounts provide safe and stable storage for your MACROMAN™ pipette controllers.
Brand:  Gilson™ F107201
 View more versions of this product
Product Code. 15891727
  • £56.30 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Keeps pipette controllers away from busy benches and desktops
  • Available in magnetic and screw varieties
  • Accessory for MACROMAN pipette controllers
  • Supplied individually
Specifications
Specifications
Screw/Adhesive Tape
Wall Mount
MACROMAN™ Pipette Controller
Black
Storage
Product Suggestions