missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Lysis Buffer T1
Lysis buffer for isolation and clean-up of genomic DNA samples using compatible kits (e.g., NucleoSpin™ Tissue kits, NucleoMag™ DNA Food).
£53.90 - £182.00
Specifications
Color | Colorless |
---|---|
Concentration | Up to 2% |
For Use With (Equipment) | Compatible kits (e.g. NucleoSpin Tissue kits, NucleoMag DNA Food) |
Formulation | Chemicals mixture in water |
pH | 7.5 to 8.5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15899137
|
Macherey-Nagel™
740940.100 |
100 mL |
£53.90
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15809147
|
Macherey-Nagel™
740940.1000 |
1000 mL |
£182.00
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Colorless | |
Compatible kits (e.g. NucleoSpin Tissue kits, NucleoMag DNA Food) | |
7.5 to 8.5 | |
Room Temperature (18°C to 25°C) | |
Isolation and cleanup of DNA |
Up to 2% | |
Chemicals mixture in water | |
Odorless | |
Liquid | |
Lysis Buffer T1 |
Safety and Handling
missing translation for 'shelfLife' : At least 1 year