Learn More
Macherey-Nagel™ Lysis Buffer T1
Lysis buffer for isolation and clean-up of genomic DNA samples using compatible kits (e.g., NucleoSpin™ Tissue kits, NucleoMag™ DNA Food).
Brand:  Macherey-Nagel™ 740940.1000
 View more versions of this product
Product Code. 15809147
  • £182.00 / 1000mL
Explore more special offers
Specifications
Colorless
Compatible kits (e.g. NucleoSpin Tissue kits, NucleoMag DNA Food)
7.5 to 8.5
Odorless
Liquid
Lysis Buffer T1
Up to 2%
Chemicals mixture in water
1000 mL
Room Temperature (18°C to 25°C)
Isolation and cleanup of DNA