missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Premium-Line Low Retention Filter Tips
Combines the advantages of standard filter tips with an innovative low retention surface to ensure maximum sample recovery, minimal sample loss and high-precision pipetting. Fisherbrand™ Premium-Line Low Retention Filter Tips eliminate sample loss and contamination.
£86.00 - £105.00
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Color | Clear |
Autoclavable | Autoclavable |
Certifications/Compliance | CE, IVD |
Product Code | Brand | Volume (Metric) | Graduated | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Graduated | Length (Metric) | Price | Quantity & Availability | |||
10264502
|
Fisherbrand™
|
0.1 to 10 μL | Yes | 34.1mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10356002
|
Fisherbrand™
|
1 to 100 μL | Yes | 50.8mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10142792
|
Fisherbrand™
|
1 to 200 μL | Yes | 50.8mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10682723
|
Fisherbrand™
|
100 to 1000 μL | Yes | 86mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10355812
|
Fisherbrand™
|
0.5 to 20 μL | No | 45.7mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Prevents sample contamination
- Made of high-quality hydrophobic virgin polyethylene without additives
- Filters block aerosols and liquids (vaporous, radioactive, biohazardous or corrosive samples) through micropore filter technology (MPS technology, filter pore size 5-40 μm)
- Optimized cone geometry provides a secure fit
- Graduation marks on tip at standard volumes provide quick and easy volume checks
- Conical tip opening prevents drop formation at the end of the tip for precise pipetting without loss of sample
- Tips made of autoclavable, non-wettable, virgin polypropylene
- Sterile version available (beta irradiation) for sterile handling
Specifications
Filtered | |
Clear | |
CE, IVD | |
Rack |
Sterile | |
Autoclavable | |
Beta radiation | |
Virgin Polypropylene |