Learn More
Fisherbrand™ Premium-Line Low Retention Filter Tips
Combines the advantages of standard filter tips with an innovative low retention surface to ensure maximum sample recovery, minimal sample loss and high-precision pipetting. Fisherbrand™ Premium-Line Low Retention Filter Tips eliminate sample loss and contamination.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 1.12500kg
Description
- Prevents sample contamination
- Made of high-quality hydrophobic virgin polyethylene without additives
- Filters block aerosols and liquids (vaporous, radioactive, biohazardous or corrosive samples) through micropore filter technology (MPS technology, filter pore size 5-40 μm)
- Optimized cone geometry provides a secure fit
- Graduation marks on tip at standard volumes provide quick and easy volume checks
- Conical tip opening prevents drop formation at the end of the tip for precise pipetting without loss of sample
- Tips made of autoclavable, non-wettable, virgin polypropylene
- Sterile version available (beta irradiation) for sterile handling
Specifications
0.5 to 20 μL | |
Sterile | |
Autoclavable | |
CE, IVD | |
Rack | |
Virgin Polypropylene |
Filtered | |
Clear | |
45.7mm | |
Beta radiation | |
No |