missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ Jackson Safety™ G40 Polyurethane Coated Gloves
£130.00
Specifications
Color Code | Black |
---|---|
Color | Black |
Material | Polyurethane |
Product Type | Gloves |
For Use With (Application) | Ideally suited use in precision assembly, electronics assembly, controls handling, packaging, and inspection |
Specifications
Black | |
Polyurethane | |
Ideally suited use in precision assembly, electronics assembly, controls handling, packaging, and inspection | |
Abrasion and tear |
Black | |
Gloves | |
60Pair | |
G40 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Kimberly-Clark™ Jackson Safety™ G40 Polyurethane Coated Gloves