missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ Jackson Safety™ G40 Polyurethane Coated Gloves
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Specifications
Specifications
Color Code | Black |
Color | Black |
Material | Polyurethane |
Product Type | Gloves |
Size | 2X-Large |
For Use With (Application) | Ideally suited use in precision assembly, electronics assembly, controls handling, packaging, and inspection |
Quantity | 60Pair |
Resistant To | Abrasion and tear |
Series | G40 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'