missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, Reagent Grade, ≥99.8% (titration), Honeywell™
Reagent Grade,≥99.8% (titration)
£19.73 - £185.38
Chemical Identifiers
CAS | 7553-56-2 |
---|---|
Molecular Formula | I2 |
Molecular Weight (g/mol) | 253.81 |
MDL Number | MFCD00011355 MFCD00164163 |
InChI Key | PNDPGZBMCMUPRI-UHFFFAOYSA-N |
Synonym | iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode |
PubChem CID | 807 |
ChEBI | CHEBI:17606 |
IUPAC Name | molecular iodine |
SMILES | II |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15627910
|
Honeywell Research Chemicals
I3380-50G |
50 g | Glass bottle | N/A | |||||
15697900
|
Honeywell Research Chemicals
I3380-100G |
100 g | Glass Bottle | N/A | |||||
15617910
|
Honeywell Research Chemicals
I3380-500G |
500 g | Glass bottle | N/A | |||||
15607910
|
Honeywell Research Chemicals
I3380-1KG |
1 kg | Glass Bottle | N/A | |||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7553-56-2 | |
253.81 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
807 | |
molecular iodine |
I2 | |
MFCD00011355 MFCD00164163 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
CHEBI:17606 | |
II |
Specifications
113.5°C | |
Black | |
I2 | |
UN3495 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
II | |
253.81 | |
CHEBI:17606 | |
≥99.8% (titration) | |
0.31mmHg (25°C), 1mmHg (38.7°C) | |
Iodine |
7553-56-2 | |
Solid | |
MFCD00011355 MFCD00164163 | |
3587194 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
molecular iodine | |
807 | |
253.81g/mol | |
Characteristic | |
Vapor Density: 9 (vs air) |
Safety and Handling
P261-P273-P280-P305 + P351 + P338-P314
missing translation for 'dotInformation' : class 8 - PG 3 - Iodine
missing translation for 'rtecsNumber' : 231-442-4