missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, Reagent Grade, ≥99.8% (titration), Honeywell™
Click to view available options
Quantity:
50 g
100 g
500 g
1 kg
Packaging:
Glass Bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Boiling Point | 184°C (literature) |
Melting Point | 113°C (literature) |
CAS Max % | 100.0000% |
Quantity | 1 kg |
Linear Formula | Fe2(SO4)3 · xH2O |
UN Number | UN3495 |
Beilstein | 3587194 |
Formula Weight | 253.81 |
Percent Purity | >99.8% |
Grade | Reagent |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'