missing translation for 'onlineSavingsMsg'
Learn More

New Pig Haz-Mat Socks

Stop the spread of acids and bases.

Brand:  New Pig HA1010

Code : Z4

Additional Details : Weight : 6.80000kg

 View more versions of this product

Product Code. 15355813

  • £134.90 / Pack of 2
Estimated Shipment: 20-05-2024
to see stock.
Add to basket

Description

Description

  • Chemical-resistant — won't degrade or cause a dangerous reaction upon contact with corrosive spills
  • Excellent for quickly diking and stopping the spread of chemical spills
  • Absorb and retain most acids, bases and unknown liquids
  • Can be incinerated after use to reduce waste

Specifications

Specifications

Pink
34.1 L
Hazmat Absorbent Sock
10 L x 5 W x 10 in. H (3.04 m x 12.7 x 2.54 cm)
10 ft.
5 in.
10 in.
5 in.
15 lb.
9 gal.
Polypropylene
Quickly diking and stopping the spread of chemical spills, absorbs acids, bases, corrosives
10 x 5 x 10 in. (3.04 m x 12.7 x 2.54 cm)
3.04 m
12.7 cm
2.54 cm
12.7 cm
6.8 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers