Learn More
Thermo Scientific™ Finnpipette™ Novus i Multichannel Electronic Pipette
Combine these pipettes with our adjustable pipette stand to assist in lower throughput MSIA sample enrichment.
£401.00 - £2215.00
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
15102589
|
Thermo Scientific™
991S |
Pipette Stand Only | N/A | N/A | N/A | |||||
15122589
|
Thermo Scientific™
991SP12 |
Pipetter, Pipette Stand | N/A | N/A | N/A | |||||
Description
Combine Thermo Scientific™ Finnpipette™ Novus i Multichannel Electronic pipettes with our adjustable pipette stand for an economical semi-automated system for rapid development and application of Mass Spectrometric Immunoassay (MSIA) methods. Each Novus i comes with a custom MSIA menu for further simplification of pipette programming that is complementary to MSIA applications.
Features:- Simplify MSIA methods using Finnpipette Novus i pipette on the pipette stand
- New software that allows 1-999 aspirate and dispense cycles with one press on the pipette trigger
- Continuous battery charging of pipette on the adjustable stand during sample processing
- Include forward, reverse, diluting, stepper pipetting, mixing, sequential stepping and more; Nine aspirate/dispense speeds
Compatible with:
Thermo Scientific MSIA D.A.R.T.'S™ (ordered separately)
Specifications
Finnpipette Novus i |