Learn More
Thermo Scientific™ Finnpipette™ Novus i Multichannel Electronic Pipette
Combine these pipettes with our adjustable pipette stand to assist in lower throughput MSIA sample enrichment.
Brand: Thermo Scientific™ 991SP12
Code : NEW
Additional Details : Weight : 2.77200kg Transport : UN number : 3481 Chem class : 9
Description
Combine Thermo Scientific™ Finnpipette™ Novus i Multichannel Electronic pipettes with our adjustable pipette stand for an economical semi-automated system for rapid development and application of Mass Spectrometric Immunoassay (MSIA) methods. Each Novus i comes with a custom MSIA menu for further simplification of pipette programming that is complementary to MSIA applications.
Features:- Simplify MSIA methods using Finnpipette Novus i pipette on the pipette stand
- New software that allows 1-999 aspirate and dispense cycles with one press on the pipette trigger
- Continuous battery charging of pipette on the adjustable stand during sample processing
- Include forward, reverse, diluting, stepper pipetting, mixing, sequential stepping and more; Nine aspirate/dispense speeds
Compatible with:
Thermo Scientific MSIA D.A.R.T.'S™ (ordered separately)
Specifications
Finnpipette Novus i electronic 12-channel Pipetter and pipette stand | |
5 to 300 μL | |
Thermo Scientific MSIA D.A.R.T.S | |
Finnpipette Novus i |