missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERMP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | ERMP1 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16807194
|
Proteintech
17321-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16897184
|
Proteintech
17321-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Within the ovary, required for the organization of somatic cells and oocytes into discrete follicular structures.Specifications
ERMP1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q3UVK0, Q6UPR8, Q7Z2K6 | |
226090, 373544, 79956 | |
ERMP1 Fusion Protein Ag11297 | |
Primary | |
-20°C | |
ERMP1 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ERMP1, Felix ina, FXNA, KIAA1815 | |
Ermp1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |