missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERMP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17321-1-AP-150UL
This item is not returnable.
View return policy
Description
Within the ovary, required for the organization of somatic cells and oocytes into discrete follicular structures.Specifications
ERMP1 | |
Polyclonal | |
Unconjugated | |
ERMP1 | |
ERMP1, Felix ina, FXNA, KIAA1815 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
226090, 373544, 79956 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3UVK0, Q6UPR8, Q7Z2K6 | |
Ermp1 | |
ERMP1 Fusion Protein Ag11297 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |