missing translation for 'onlineSavingsMsg'
Learn More

DL-Malic acid, +99%, Thermo Scientific Chemicals

£20.00 - £166.83

Chemical Identifiers

CAS 6915-15-7
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.087
MDL Number MFCD00064212
InChI Key BJEPYKJPYRNKOW-UHFFFAOYSA-N
Synonym malic acid, dl-malic acid, 2-hydroxysuccinic acid, malate, hydroxysuccinic acid, butanedioic acid, hydroxy, kyselina jablecna, pomalus acid, hydroxybutanedioic acid, deoxytetraric acid
PubChem CID 525
ChEBI CHEBI:6650
IUPAC Name 2-hydroxybutanedioic acid
SMILES C(C(C(=O)O)O)C(=O)O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10256523
View Documents
Thermo Scientific Acros
125252500
250 g Plastic bottle
£20.00
250g
Please to purchase this item. Need a web account? Register with us today!
10025723
View Documents
Thermo Scientific Acros
125255000
500 g Plastic bottle
£31.76
500g
Please to purchase this item. Need a web account? Register with us today!
10584051
View Documents
Thermo Scientific Acros
125250010
1 kg Plastic bottle
£51.66
1kg
Please to purchase this item. Need a web account? Register with us today!
11965071
View Documents
Thermo Scientific Acros
125250050
5 kg Plastic Drum
£166.83
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6915-15-7
134.087
BJEPYKJPYRNKOW-UHFFFAOYSA-N
525
2-hydroxybutanedioic acid
C4H6O5
MFCD00064212
malic acid, dl-malic acid, 2-hydroxysuccinic acid, malate, hydroxysuccinic acid, butanedioic acid, hydroxy, kyselina jablecna, pomalus acid, hydroxybutanedioic acid, deoxytetraric acid
CHEBI:6650
C(C(C(=O)O)O)C(=O)O

Specifications

6915-15-7
99+%
HO2CCH2CH(OH)CO2H
malic acid, dl-malic acid, 2-hydroxysuccinic acid, malate, hydroxysuccinic acid, butanedioic acid, hydroxy, kyselina jablecna, pomalus acid, hydroxybutanedioic acid, deoxytetraric acid
BJEPYKJPYRNKOW-UHFFFAOYSA-N
2-hydroxybutanedioic acid
525
134.09
203°C
128.0°C to 132.0°C
C4H6O5
MFCD00064212
Solubility in water: 558g/L (20°C). Other solubilities: 82.70 g/100 g methanol-17.75 g/100 g acetone (20°C, 45.53 g/100 g ethanol-20.70 g/ 100 g dioxane (20°C, 0.84 g/100 g diethyl ether (20°C), practically insoluble in benzene
C(C(C(=O)O)O)C(=O)O
134.087
CHEBI:6650
99+%
DL-Malic acid
Videos
Safety and Handling

Safety and Handling

GHS P Statement Causes serious eye irritation.

GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger

Product Identifier
  • DL-Malic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.