missing translation for 'onlineSavingsMsg'
Learn More

DL-Malic acid, +99%, Thermo Scientific Chemicals

Product Code. 11965071
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6915-15-7
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.087
MDL Number MFCD00064212
InChI Key BJEPYKJPYRNKOW-UHFFFAOYSA-N
Synonym malic acid, dl-malic acid, 2-hydroxysuccinic acid, malate, hydroxysuccinic acid, butanedioic acid, hydroxy, kyselina jablecna, pomalus acid, hydroxybutanedioic acid, deoxytetraric acid
PubChem CID 525
ChEBI CHEBI:6650
IUPAC Name 2-hydroxybutanedioic acid
SMILES C(C(C(=O)O)O)C(=O)O

Specifications

Melting Point 128.0°C to 132.0°C
Assay Percent Range 99+%
Linear Formula HO2CCH2CH(OH)CO2H
Solubility Information Solubility in water: 558g/L (20°C). Other solubilities: 82.70 g/100 g methanol-17.75 g/100 g acetone (20°C, 45.53 g/100 g ethanol-20.70 g/ 100 g dioxane (20°C, 0.84 g/100 g diethyl ether (20°C), practically insoluble in benzene
Formula Weight 134.09
Percent Purity 99+%
Quantity 5 kg
Flash Point 203°C
Packaging Plastic Drum
Chemical Name or Material DL-Malic acid
compliance-icons
Product Identifier
  • DL-Malic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.