missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexene-1-boronic acid, 97%, Thermo Scientific™
£108.00 - £320.00
Chemical Identifiers
CAS | 89490-05-1 |
---|---|
Molecular Formula | C6H11BO2 |
Molecular Weight (g/mol) | 125.96 |
MDL Number | MFCD02179497 |
InChI Key | XZWQKJXJNKYMAP-UHFFFAOYSA-N |
Synonym | cyclohex-1-en-1-ylboronic acid, 1-cyclohexenylboronic acid, 1-cyclohexen-1-yl-boronic acid, cyclohexenylboronic acid, cyclohexen-1-yl-boronic acid, 1-cyclohexen-1-ylboronic acid, cyclohexene-1-boronic acid, 1-boronocyclohex-1-ene, cyclohex-1-enylboronic acid, cyclohexen-1-yl boronic acid |
PubChem CID | 3862902 |
IUPAC Name | cyclohexen-1-ylboronic acid |
SMILES | OB(O)C1=CCCCC1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
89490-05-1 | |
125.96 | |
XZWQKJXJNKYMAP-UHFFFAOYSA-N | |
3862902 | |
OB(O)C1=CCCCC1 |
C6H11BO2 | |
MFCD02179497 | |
cyclohex-1-en-1-ylboronic acid, 1-cyclohexenylboronic acid, 1-cyclohexen-1-yl-boronic acid, cyclohexenylboronic acid, cyclohexen-1-yl-boronic acid, 1-cyclohexen-1-ylboronic acid, cyclohexene-1-boronic acid, 1-boronocyclohex-1-ene, cyclohex-1-enylboronic acid, cyclohexen-1-yl boronic acid | |
cyclohexen-1-ylboronic acid |
Specifications
89490-05-1 | |
C6H11BO2 | |
cyclohex-1-en-1-ylboronic acid, 1-cyclohexenylboronic acid, 1-cyclohexen-1-yl-boronic acid, cyclohexenylboronic acid, cyclohexen-1-yl-boronic acid, 1-cyclohexen-1-ylboronic acid, cyclohexene-1-boronic acid, 1-boronocyclohex-1-ene, cyclohex-1-enylboronic acid, cyclohexen-1-yl boronic acid | |
OB(O)C1=CCCCC1 | |
125.96 | |
125.96 | |
Cyclohexene-1-boronic acid |
165°C | |
MFCD02179497 | |
XZWQKJXJNKYMAP-UHFFFAOYSA-N | |
cyclohexen-1-ylboronic acid | |
3862902 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C