missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexene-1-boronic acid, 97%, Thermo Scientific™

Product Code. 15450008
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89490-05-1
Molecular Formula C6H11BO2
Molecular Weight (g/mol) 125.96
MDL Number MFCD02179497
InChI Key XZWQKJXJNKYMAP-UHFFFAOYSA-N
Synonym cyclohex-1-en-1-ylboronic acid, 1-cyclohexenylboronic acid, 1-cyclohexen-1-yl-boronic acid, cyclohexenylboronic acid, cyclohexen-1-yl-boronic acid, 1-cyclohexen-1-ylboronic acid, cyclohexene-1-boronic acid, 1-boronocyclohex-1-ene, cyclohex-1-enylboronic acid, cyclohexen-1-yl boronic acid
PubChem CID 3862902
IUPAC Name cyclohexen-1-ylboronic acid
SMILES OB(O)C1=CCCCC1

Specifications

Melting Point 165°C
Quantity 1 g
Formula Weight 125.96
Percent Purity 97%
Chemical Name or Material Cyclohexene-1-boronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'