Learn More
Chloroacetyl chloride, 98%, Thermo Scientific Chemicals
£17.10 - £132.00
Chemical Identifiers
CAS | 79-04-9 |
---|---|
Molecular Formula | C2H2Cl2O |
Molecular Weight (g/mol) | 112.937 |
MDL Number | MFCD00000725 |
InChI Key | VGCXGMAHQTYDJK-UHFFFAOYSA-N |
Synonym | chloroacetyl chloride, acetyl chloride, chloro, chloracetyl chloride, chloroacetic chloride, monochloroacetyl chloride, chloroacetic acid chloride, chloroacetylchloride, chlorure de chloracetyle, unii-k5uml06yuo, chlorid kyseliny chloroctove |
PubChem CID | 6577 |
ChEBI | CHEBI:34624 |
IUPAC Name | 2-chloroacetyl chloride |
SMILES | C(C(=O)Cl)Cl |
Description
Chloroacetyl chloride is an intermediate in the production of alachlor and butachlor. It is used to prepare phenacyl chloride through Friedel-Crafts acylation of benzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChloroacetyl chloride is an intermediate in the production of alachlor and butachlor. It is used to prepare phenacyl chloride through Friedel-Crafts acylation of benzene.
Solubility
Miscible with ethyl ether, acetone, benzene and carbon tetrachloride.
Notes
Incompatible with strong oxidizing agents, strong bases, alcohols and water. Moisture sensitive.
Chemical Identifiers
79-04-9 | |
112.937 | |
VGCXGMAHQTYDJK-UHFFFAOYSA-N | |
6577 | |
2-chloroacetyl chloride |
C2H2Cl2O | |
MFCD00000725 | |
chloroacetyl chloride, acetyl chloride, chloro, chloracetyl chloride, chloroacetic chloride, monochloroacetyl chloride, chloroacetic acid chloride, chloroacetylchloride, chlorure de chloracetyle, unii-k5uml06yuo, chlorid kyseliny chloroctove | |
CHEBI:34624 | |
C(C(=O)Cl)Cl |
Specifications
79-04-9 | |
1.417 | |
Pungent | |
ClCH2COCl | |
MFCD00000725 | |
605439 | |
14,2067 | |
Miscible with ethyl ether,acetone,benzene and carbon tetrachloride. | |
C(C(=O)Cl)Cl | |
112.937 | |
CHEBI:34624 | |
98% |
-22°C | |
105°C to 106°C | |
C2H2Cl2O | |
1.453 | |
UN1752 | |
Moisture sensitive | |
chloroacetyl chloride, acetyl chloride, chloro, chloracetyl chloride, chloroacetic chloride, monochloroacetyl chloride, chloroacetic acid chloride, chloroacetylchloride, chlorure de chloracetyle, unii-k5uml06yuo, chlorid kyseliny chloroctove | |
VGCXGMAHQTYDJK-UHFFFAOYSA-N | |
2-chloroacetyl chloride | |
6577 | |
112.94 | |
Chloroacetyl chloride |
Safety and Handling
GHS H Statement
H301-H311-H331-H372-H314
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
H290-H301+H311+H331-H314-H335-H372
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: CHLOROACETYL CHLORIDE
missing translation for 'einecsNumber' : 201-171-6
missing translation for 'rtecsNumber' : AO6475000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.