missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11600222
Click to view available options
Quantity:
25 mL
250 mL
500 mL
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Chloroacetyl chloride is an intermediate in the production of alachlor and butachlor. It is used to prepare phenacyl chloride through Friedel-Crafts acylation of benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetyl chloride is an intermediate in the production of alachlor and butachlor. It is used to prepare phenacyl chloride through Friedel-Crafts acylation of benzene.

Solubility
Miscible with ethyl ether, acetone, benzene and carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents, strong bases, alcohols and water. Moisture sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-04-9
Molecular Formula C2H2Cl2O
Molecular Weight (g/mol) 112.937
MDL Number MFCD00000725
InChI Key VGCXGMAHQTYDJK-UHFFFAOYSA-N
Synonym chloroacetyl chloride, acetyl chloride, chloro, chloracetyl chloride, chloroacetic chloride, monochloroacetyl chloride, chloroacetic acid chloride, chloroacetylchloride, chlorure de chloracetyle, unii-k5uml06yuo, chlorid kyseliny chloroctove
PubChem CID 6577
ChEBI CHEBI:34624
IUPAC Name 2-chloroacetyl chloride
SMILES C(C(=O)Cl)Cl

Specifications

Melting Point -22°C
Density 1.417
Boiling Point 105°C to 106°C
Odor Pungent
Linear Formula ClCH2COCl
Refractive Index 1.453
Quantity 500 mL
UN Number UN1752
Beilstein 605439
Sensitivity Moisture sensitive
Merck Index 14,2067
Solubility Information Miscible with ethyl ether,acetone,benzene and carbon tetrachloride.
Formula Weight 112.94
Percent Purity 98%
Chemical Name or Material Chloroacetyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.