Learn More
Astell Scientific™ Category 3 (BSL-3) Compliance Option
Required when dealing with high risk sterilization requirements. Astell Scientific™ Category 3 (BSL-3) Compliance Option uses a bacterial retentive filter fitted onto the exhaust to ensure that nothing leaves the chamber without being sterilized.
£1567.09 - £3030.00
Specifications
Certifications/Compliance | Category 3 (BSL-3) |
---|---|
For Use With (Application) | High-Risk Sterilization Processes |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15721459
|
Astell Scientific™
AVQ005 |
33, 43, 63 L Compact Autoclaves | N/A | ||||||
15731459
|
Astell Scientific™
AVQ006 |
Benchtop Autoclave Range, Compact Autoclave Range, Top Loading Autoclave Range, Front Loading Autoclave Range, Duaclave Autoclave Range, SQUARE ECO Autoclave Range, SQUARE Autoclave Range, Double Door SQUARE Autoclave Range | N/A | ||||||
Description
- Required for high risk sterilization processes.
- Bacterial retentive filter.
- Prevents anything from leaving the autoclave chamber without being sterilized.
Specifications
Category 3 (BSL-3) | |
High-Risk Sterilization Processes |