Learn More
Astell Scientific™ Category 3 (BSL-3) Compliance Option
Required when dealing with high risk sterilization requirements. Astell Scientific™ Category 3 (BSL-3) Compliance Option uses a bacterial retentive filter fitted onto the exhaust to ensure that nothing leaves the chamber without being sterilized.
Brand: Astell Scientific™ AVQ006
Additional Details : Weight : 0.01000kg
Description
- Required for high risk sterilization processes.
- Bacterial retentive filter.
- Prevents anything from leaving the autoclave chamber without being sterilized.
Specifications
Category 3 (BSL-3) | |
High-Risk Sterilization Processes | |
Benchtop Autoclave Range, Compact Autoclave Range, Top Loading Autoclave Range, Front Loading Autoclave Range, Duaclave Autoclave Range, SQUARE ECO Autoclave Range, SQUARE Autoclave Range, Double Door SQUARE Autoclave Range |