missing translation for 'onlineSavingsMsg'
Learn More
Buffer, Mercury Free, 25°C, Reagecon™ Product Code.: 12313669

Buffer, Mercury Free, 25°C, Reagecon™

Product Code. 12313669 Shop All Reagecon Products
5 L, Red, 4 ±0.01
Click to view available options
Quantity:
1 L
5 L
Color:
Clear
Red
Yellow
pH:
4 ±0.01
7 ±0.01
This item is not returnable. View return policy

Product Code. 12313669

Brand: Reagecon 5040MF

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used with pH meters to fit pH of the reaction media. Reagecon Buffer, Mercury Free, 25°C are colored buffer solutions with an extended shelf life. Available in pH 4 and 10.

pH values are certified at 25°C to maximize confidence in pH measurements. As well as being ideal for use as calibration standards, they are also cover values that can be used as control standards for all pH applications.

  • Mercury free, color-coded pH buffers at 25°C
  • Extended shelf life
  • Supplied with a detailed certificate of analysis, which outlines traceability to NIST
  • Temperature dependence data is printed on label, as are lot numbers and expiry dates

Specifications

Color Red
Certificate of Analysis Supplied
pH 4 ±0.01
Physical Form Liquid
Quantity 5 L
Packaging Bottle
Traceability to NIST Traceable to NIST
Solubility Information Fully miscible
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer, Mercury Free, 25°C, Reagecon™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.