missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 4.00 Buffer Solution at 25°C, Reagecon
Tested, certified, NIST traceable, and shelf-stable buffer solution.
Brand: Reagecon 504025
This item is not returnable.
View return policy
Description
- pH Buffer Reference Standard, pH Buffer Reference Material
- Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Clear | |
4.00 ±0.01 | |
5 L |
Accuracy ± 0.01 pH units | |
Liquid | |
HDPE Bottle GL45 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction