Learn More
Biphenyl-4-boronic acid, 97+%, Thermo Scientific Chemicals
£23.70 - £260.00
Chemical Identifiers
CAS | 5122-94-1 |
---|---|
Molecular Formula | C12H11BO2 |
Molecular Weight (g/mol) | 198.03 |
MDL Number | MFCD00093311 |
InChI Key | XPEIJWZLPWNNOK-UHFFFAOYSA-N |
Synonym | 4-biphenylboronic acid, 1,1'-biphenyl-4-ylboronic acid, biphenyl-4-boronic acid, 4-biphenyl boronic acid, biphenylboronic acid, 4-phenylphenyl boranediol, 4-phenylphenyl boronic acid, biphenyl-4-ylboronic acid, 1,1'-biphenyl-4-yl-boronic acid, 4-phenylbenzeneboronic acid |
PubChem CID | 151253 |
IUPAC Name | (4-phenylphenyl)boronic acid |
SMILES | OB(O)C1=CC=C(C=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11480118
|
Thermo Scientific Alfa Aesar
B23703.03 |
1 g |
£23.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490118
|
Thermo Scientific Alfa Aesar
B23703.06 |
5 g |
£82.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400128
|
Thermo Scientific Alfa Aesar
B23703.14 |
25 g |
£260.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used in Suzuki reactions and as intermediates of liquid crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed in Suzuki reactions and as intermediates of liquid crystals.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
5122-94-1 | |
198.03 | |
XPEIJWZLPWNNOK-UHFFFAOYSA-N | |
151253 | |
OB(O)C1=CC=C(C=C1)C1=CC=CC=C1 |
C12H11BO2 | |
MFCD00093311 | |
4-biphenylboronic acid, 1,1'-biphenyl-4-ylboronic acid, biphenyl-4-boronic acid, 4-biphenyl boronic acid, biphenylboronic acid, 4-phenylphenyl boranediol, 4-phenylphenyl boronic acid, biphenyl-4-ylboronic acid, 1,1'-biphenyl-4-yl-boronic acid, 4-phenylbenzeneboronic acid | |
(4-phenylphenyl)boronic acid |
Specifications
5122-94-1 | |
Odorless | |
MFCD00093311 | |
4-biphenylboronic acid, 1,1'-biphenyl-4-ylboronic acid, biphenyl-4-boronic acid, 4-biphenyl boronic acid, biphenylboronic acid, 4-phenylphenyl boranediol, 4-phenylphenyl boronic acid, biphenyl-4-ylboronic acid, 1,1'-biphenyl-4-yl-boronic acid, 4-phenylbenzeneboronic acid | |
XPEIJWZLPWNNOK-UHFFFAOYSA-N | |
(4-phenylphenyl)boronic acid | |
151253 | |
≥98% |
238°C to 247°C | |
C12H11BO2 | |
2937410 | |
Insoluble in water. | |
OB(O)C1=CC=C(C=C1)C1=CC=CC=C1 | |
198.03 | |
198.03 | |
Biphenyl-4-boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only