missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-4-boronic acid, 97+%, Thermo Scientific Chemicals

Product Code. 11400128
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in Suzuki reactions and as intermediates of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in Suzuki reactions and as intermediates of liquid crystals.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5122-94-1
Molecular Formula C12H11BO2
Molecular Weight (g/mol) 198.03
MDL Number MFCD00093311
InChI Key XPEIJWZLPWNNOK-UHFFFAOYSA-N
Synonym 4-biphenylboronic acid, 1,1'-biphenyl-4-ylboronic acid, biphenyl-4-boronic acid, 4-biphenyl boronic acid, biphenylboronic acid, 4-phenylphenyl boranediol, 4-phenylphenyl boronic acid, biphenyl-4-ylboronic acid, 1,1'-biphenyl-4-yl-boronic acid, 4-phenylbenzeneboronic acid
PubChem CID 151253
IUPAC Name (4-phenylphenyl)boronic acid
SMILES OB(O)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 238°C to 247°C
Odor Odorless
Quantity 25 g
Beilstein 2937410
Solubility Information Insoluble in water.
Formula Weight 198.03
Percent Purity ≥98%
Chemical Name or Material Biphenyl-4-boronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'