Learn More
Sampling Systems™ Angular Beaker
These angular beaker attachments allow the cup to be set in specific locations while using the TeleScoop.
Brand: Sampling Systems™ 5065B-1001B
This item is not returnable.
View return policy
Description
- Telescopic samplers are often used for taking surface samples from rivers, tanks and lakes
- A wide choice of attachments is available to make the TeleScoop ideally suited for most applications
- Attachments are interchangeable and simply click into position
Specifications
Swabbing Template | |
Autoclavable | |
Reusable | |
Non-sterile |
Stainless Steel | |
1 L | |
No |