missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-pentanone, puriss. p.a., ACS Reagent, ≥99%, Honeywell Riedel-de Haën™
Puriss. p.a., ACS Reagent, Reag. Ph. Eur., for extraction analysis,≥99%
£328.38 - £479.49
Chemical Identifiers
CAS | 108-10-1 |
---|---|
Molecular Formula | C6H12O |
Molecular Weight (g/mol) | 100.161 |
MDL Number | MFCD00008938 |
InChI Key | NTIZESTWPVYFNL-UHFFFAOYSA-N |
Synonym | 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon |
PubChem CID | 7909 |
ChEBI | CHEBI:82344 |
IUPAC Name | 4-methylpentan-2-one |
SMILES | CC(C)CC(=O)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15663980
|
Honeywell-Riedel-de Haen
33463-6X1L |
6 x 1 L | Glass Bottle |
£328.38
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15653980
|
Honeywell-Riedel-de Haen
33463-4X2.5L |
4 x 2.5 L | Glass Bottle |
£479.49
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
108-10-1 | |
100.161 | |
NTIZESTWPVYFNL-UHFFFAOYSA-N | |
7909 | |
4-methylpentan-2-one |
C6H12O | |
MFCD00008938 | |
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon | |
CHEBI:82344 | |
CC(C)CC(=O)C |
Specifications
108-10-1 | |
Colorless | |
117°C | |
Characteristic | |
(CH3)2CHCH2COCH3 | |
UN1245 | |
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon | |
CC(C)CC(=O)C | |
100.161 | |
CHEBI:82344 | |
≥99% | |
4-Methyl-2-pentanone, For extraction analysis |
-80°C | |
0.801g/mL at 25°C | |
14°C (57.2°F) | |
C6H12O | |
MFCD00008938 | |
605399 | |
NTIZESTWPVYFNL-UHFFFAOYSA-N | |
4-methylpentan-2-one | |
7909 | |
100.16g/mol | |
ACS/Reagent Ph. Eur./puriss. p.a. |
Safety and Handling
P210-P261-P305 + P351 + P338
missing translation for 'dotInformation' : Class 3 - PG 2 - Methyl isobutyl ketone
missing translation for 'rtecsNumber' : 203-550-1