missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone, puriss. p.a., ACS Reagent, ≥99%, Honeywell Riedel-de Haën™

Product Code. 15653980
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. Ph. Eur., for extraction analysis,≥99%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00008938
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
Synonym 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
PubChem CID 7909
ChEBI CHEBI:82344
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(=O)C

Specifications

Melting Point -80°C
Color Colorless
Density 0.801g/mL at 25°C
Boiling Point 117°C
Flash Point 14°C (57.2°F)
Odor Characteristic
Packaging Glass Bottle
Linear Formula (CH3)2CHCH2COCH3
Quantity 4 x 2.5 L
UN Number UN1245
Beilstein 605399
Formula Weight 100.16g/mol
Percent Purity ≥99%
Grade ACS/Reagent Ph. Eur./puriss. p.a.
Chemical Name or Material 4-Methyl-2-pentanone, For extraction analysis
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'